Learn More
PRELID1 Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811047
Description
May be important for the development of vital and immunocompetent organs.Specifications
| PRELID1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y255 | |
| PRELID1 | |
| Human recombit protein fragment corresponding to amino acids 36-219 of human PRELID1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A6 | |
| Unconjugated | |
| PRELID1 | |
| CGI-106, PRELI, PX19, SBBI12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27166 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.