Learn More
Prednisolone hemisuccinate, MedChemExpress
Product Code. 16498596
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16498596
Brand: Medchem Express HYB1087
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Prednisolone hemisuccinate is a synthetic glucocorticoid, a derivative of cortisol, which is used to treat a variety of inflammatory and auto-immune conditions.
Prednisolone hemisuccinate is a synthetic glucocorticoid, a derivative of cortisol, which is used to treat a variety of inflammatory and auto-immune conditions.
Specifications
Chemical Name or Material Prednisolone hemisuccinate
CAS 2920-86-7
Color White
Purity Grade Notes Research
Molecular Formula C25H32O8
Quantity 100 mg
Synonym Prednisolone 21-hemisuccinate
Solubility Information DMSO : 250 mg/mL (542.86 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C(COC(CCC(O)=O)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 460.52
Formula Weight 460.52
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation