missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Pre-Assembled Screw Top Vial and Closure Combination Pack N24
Shop All Macherey-Nagel ProductsDescription
- Pre-sealed vials and caps reduce contamination risk
- 1st hydrolytic class clear glass vials
- Caps with hole and pre-fitted red rubber/PTFE septa
- 45° shore A
- Bonded assembly
Specifications
Specifications
| Closure Color | White |
| Color (Seal) | Red-Orange/Colorless |
| Description | N24 Screw Top Vial and Pre-Sealed Cap Combination Pack |
| Color | Clear |
| Material | Hydrolytic Glass (Class I) |
| Septum | Red Rubber/PTFE |
| Neck Style | Screw Neck |
| Base Style | Flat Bottom |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap, Center Hole |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?