missing translation for 'onlineSavingsMsg'
Learn More

PRE-084 hydrochloride, MedChemExpress

Product Code. p-200102010
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16473487

Brand: Medchem Express HY18100A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. PRE-084 hydrochloride exhibits good neuroprotective effects, can improve motor function and motor neuron survival in mice. PRE-084 hydrochloride also can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.

PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. PRE-084 hydrochloride exhibits good neuroprotective effects, can improve motor function and motor neuron survival in mice. PRE-084 hydrochloride also can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.

Specifications

Chemical Name or Material PRE-084 hydrochloride
Physical Form Solid
CAS 75136-54-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H28ClNO3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 34 mg/mL (96.08 mM) ∣H2O : 33.33 mg/mL (94.18 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1(C2=CC=CC=C2)CCCCC1)OCCN3CCOCC3.[H]Cl
Molecular Weight (g/mol) 353.88
Formula Weight 353.88
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.