Learn More
PRE-084 hydrochloride, MedChemExpress
Description
PRE-084 hydrochloride is a highly selective σ1 receptor (S1R) agonist, with an IC50 of 44 nM. PRE-084 hydrochloride exhibits good neuroprotective effects, can improve motor function and motor neuron survival in mice. PRE-084 hydrochloride also can ameliorate myocardial ischemia-reperfusion injury in rats by activating the Akt-eNOS pathway.
Specifications
Specifications
| Chemical Name or Material | PRE-084 hydrochloride |
| Physical Form | Solid |
| CAS | 75136-54-8 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H28ClNO3 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : ≥ 34 mg/mL (96.08 mM) ∣H2O : 33.33 mg/mL (94.18 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1(C2=CC=CC=C2)CCCCC1)OCCN3CCOCC3.[H]Cl |
| Show More |