Learn More
PRDX6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-13585-20UL
 View more versions of this product
Product Code. 18074785
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The PRDX6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRDX6. This antibody recognizes human, mouse, rat antigen. The PRDX6 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
PRDX6
1000 μg/mL
1 Cys, 1 Cys peroxiredoxin, 1 Cys PRX, 24 kDa protein, aiPLA2, Antioxidant protein 2, AOP2, KIAA0106, Liver 2D page spot 40, NSGPx, p29, peroxiredoxin 6, PRDX6, PRX, Red blood cells page spot 12
20 μL
9588
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa