missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Prasugrel

Product Code. 18724089
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18724089 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724089 Supplier Tocris Bioscience™ Supplier No. 6317/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversible P2Y12 antagonist; antiplatelet and orally active

Prasugrel is an irreversible P2Y12 antagonist; metabolised to active metabolite R-99224 (IC50 = 45 μM) irreversibly binds platelet P2Y12 receptors, inhibiting platelet activation. Antiplatelet and antithrombotic. Orally active and active in vivo. Note - inactive in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(2-fluorophenyl)ethanone
CAS 150322-43-3
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C20H20FNO3S
Formula Weight Observed MW: 373.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.