Learn More
Pranlukast, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Pranlukast |
| CAS | 103177-37-3 |
| Molecular Formula | C27 H23 N5 O4 |
| Quantity | 250 mg |
| Synonym | Benzamide','N-[4-oxo-2-(2H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)-, Benzamide','N-[4-oxo-2-(1H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)- (9CI), N-[4-Oxo-2-(2H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)benzamide, 4-Oxo-8-[4-(4-phenylbutoxy)benzoylamino]-2-(tetrazol-5-yl)-4H-1-benzopyran, 8-[4-(4-Phenylbutoxy)benzoyl]amino-2-(5-tetrazolyl)-4-oxo-4H-1-benzopyran, Azlaire, CCN 00401, ONO 1078, ONO-RS 411, Onon, Pranlukast, RS 411, SB 205312 |
| InChI Formula | InChI=1S/C27H23N5O4/c33-23-17-24(26-29-31-32-30-26)36-25-21(23)10-6-11-22(25)28-27(34)19-12-14-20(15-13-19)35-16-5-4-9-18-7-2-1-3-8-18/h1-3,6-8,10-15,17H,4-5,9,16H2,(H,28,34)(H,29,30,31,32) |
| SMILES | O=C(Nc1cccc2C(=O)C=C(Oc12)c3nnn[nH]3)c4ccc(OCCCCc5ccccc5)cc4 |
| IUPAC Name | N-[4-oxo-2-(1H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide |
| Molecular Weight (g/mol) | 481.5 |
| Formula Weight | 481.175 |