Learn More
Eurofins DiscoverX PRAK Protein, Active
Description
Protein Serine/Threonine Kinase MAPKAPK5 (mitogen-activated protein kinase activated protein kinase 5), also known as p38-regulated/activated protein kinase (PRAK), functions downstream of p38a and p38b in mediating signaling of the p38 pathway. This kinase can be activated in response to cellular stress and proinflammatory cytokines through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. MAPKAPK5 is localized predominantly in the cytoplasm and is constantly shuttled between the cytoplasm and the nucleus. MAPKAPK5 may play a role in balancing other MAPK pathways because overactivation of MAPKAPK5 can inhibit Ras mediated cell proliferation and gene activation. MAPKAPK5 inhibtion may facilitate cell differentiation, inflammation and cell proliferation.
EMBL AF032437
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 270 mM sucrose. Frozen solution. |
| Molecular Weight (g/mol) | 55 kDa |
| Quantity | 15 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥57% |
| Protein | PRAK, active |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.