missing translation for 'onlineSavingsMsg'
Learn More

Praeruptorin B, MedChemExpress

Product Code. 16433836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16433836

Brand: Medchem Express HYN0082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Praeruptorin B is an inhibitor of sterol regulatory element-binding proteins (SREBPs).

Praeruptorin B is an inhibitor of sterol regulatory element-binding proteins (SREBPs).

Specifications

Chemical Name or Material Praeruptorin B
CAS 73069-28-0
Color White
Purity Grade Notes Research
Molecular Formula C24H26O7
Quantity 25 mg
Synonym Praeruptorin D
Solubility Information DMSO : 25 mg/mL (58.62 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C=CC2=CC=C3C([C@H](OC(/C(C)=C\C)=O)[C@H](OC(/C(C)=C\C)=O)C(C)(C)O3)=C2O1
Molecular Weight (g/mol) 426.46
Formula Weight 426.46
Percent Purity 99.88%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.