Learn More
Bohlender™ PPS Screw Caps With Aperture
Product Code. 10296883
Click to view available options
Thread Size:
GL 18
GL 25
Unit Size:
Pack of 10
This item is not returnable. View return policy
Product Code. 10296883
Brand: Bohlender™ H99518
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Use these caps as a joining piece between tubes with flange and tubes with GL thread.
  • PBTP Screw cap with handy knurl
  • Suitable for GL threads
  • With aperture
  • Use as septum by inserting a separately available gasket (Cat. No. H 973-..)
  • Use as a joining piece between tubes with flange and tubes with GL thread
  • Suitable gasket rings are available separately (Cat. No. H 975-.. / H 977-..)
Specifications
Certifications/Compliance FDA Conform
Material PPS
For Use With For GL 18 thread
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 1 Each
Thread Size GL 18
Max. Operating Temperature (Metric) 250°C
Product Type Screw Cap