missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R14D Polyclonal Antibody
GREENER_CHOICE

Product Code. 16688254
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16688254 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16688254 Supplier Invitrogen™ Supplier No. PA5101783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP1R14D.

Protein phosphatase-1 (PP1; see MIM 176875) is a major cellular phosphatase that reverses serine/threonine protein phosphorylation. PPP1R14D is a PP1 inhibitor that itself is regulated by phosphorylation (Liu et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R14D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R14D
Gene Accession No. Q9NXH3
Gene Alias 2010107K19Rik; CPI17-like; gastrointestinal and brain-specific PP1-inhibitory protein 1; GBPI; GBPI1; GBPI-1; gut and brain phosphatase inhibitor 1; PKC-dependent PP1 inhibitory protein; PKC-dependent PP1 inhibitory protein subunit 14; Ppp1r14; PPP1R14D; protein phosphatase 1 regulatory inhibitor subunit 14D; protein phosphatase 1 regulatory subunit 14D; protein phosphatase 1, regulatory (inhibitor) subunit 14D
Gene Symbols PPP1R14D
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R14D(Accession Q9NXH3), corresponding to amino acid residues Q65-I115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54866
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.