Learn More
Bohlender™ PP Blue Multiple Distributors for Bottles
Description
- Providing temperature resistance from 0° to +110°C
- Very good chemical resistance
- FDA conformity
- Vacuum suitable
- Autoclavable at 121°C
- Blue screw cap made of PP with GLS 80 thread, body made of PP
- 4 necks with GL 18 thread
- For use with tubing of max. O.D. 4 x 12.7 mm
Specifications
Specifications
| Certifications/Compliance | FDA |
| Description | 4 x GL 18 necks |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Tubing max. O.D. 4 x 12.7 mm |
| For Use With (Application) | Inserting/drawing liquids, probes, tubing |
| Min. Operating Temperature (Metric) | 0°C |
| Resistance Properties | Very good chemical resistance |
| Thread Size | GL 18 |
| Color | Blue |
| Show More |