missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU2F3, Mouse anti-Human, Clone: 6D1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00025833-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MVNLESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQSpecifications
| POU2F3 | |
| Monoclonal | |
| Unconjugated | |
| POU class 2 homeobox 3 | |
| Epoc-1/FLJ40063/MGC126698/OCT11/PLA-1/Skn-1a | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6D1 | |
| In 1x PBS, pH 7.4 | |
| NM_014352 | |
| POU2F3 | |
| POU2F3 (NP_055167, 1 a.a. ∼ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 25833 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction