Learn More
Potassium Hydroxide, 1M, Chem Lab™
Product Code. 11963173
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11963173 1 L 1 L
11973173 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11963173 Supplier Chem-Lab Supplier No. CL05.1108.1000
 Restricted Item
In Stock
Add to basket
Add to basket
Specifications
pH >13
Color Colorless
Concentration 56.11g KOH/l H2O = 1 N (±0.002/20°C
Density 1.05g/mL
Solubility Information Soluble in Water
Quantity 1 L
Odor Odorless
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 1 mol/l
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide