missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, Free-flowing, ACS reagent, anhydrous, ≥99.0%, Honeywell Fluka™

Product Code. 15657080 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15657080

Brand: Honeywell-Fluka 791733100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 100 g
Linear Formula CH3COOK
Beilstein 3595449
Formula Weight 98.14
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.57 g/cm3 at 25 °C(lit.)
Assay titration with perchloric acid
Chemical Name or Material Potassium acetate, Free-flowing, ACS reagent, anhydrous, ≥99.0%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.