missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Portable Support Clamp

Product Code. 15752439 Shop All Ohaus Products
Click to view available options
Material:
Cast Aluminum
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15752439

Brand: Ohaus™ 30400023

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

OHAUS™ Portable Support Clamp provides secure support for holding gas cylinders at elevated positions.

  • Suitable for holding glassware safely
  • For portable applications
  • Constructed of aluminum
TRUSTED_SUSTAINABILITY

Specifications

Type Floor Stand
Material Cast Aluminum
For Use With (Application) Cas Cylinder Support
Description Clamp, Support, 713 Portable, CLR-713
Diameter (Metric) 394.00 mm
Height (Metric) 76.000 mm
Height (English) 3.00 in.
Width (Metric) 394 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.