missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Analog pH/mV/°C Meter with Electrodes

Product Code. 10478253
Click to view available options
Dimensions (L x W x H):
145 x 80 x 36 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10478253

Brand: Hanna Instruments™ HI83141

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to be accurate, reliable and easy to use. Hanna Instruments™ Analog pH/mV/°C Meter with Electrodes is a portable multiparameter meter that uses the HI1230B pH electrode and HI7669AW temperature probe (both included) using separate connections.

Manual calibration is performed at one or two points by adjusting the trimmers on the front panel. Capable of measuring pH/mV and °C, this meter is great for field work, providing one meter for multiple uses. This instrument is ideal for applications that require a custom calibration point. Manual calibration can be extremely useful in order to achieve better accuracy.

  • Automatic Temperature Compensation (ATC)
  • Two-point Calibration
  • Compact, heavy-duty, and waterproof protected casing
  • Temperature probe and pH electrode included
  • Can also be used for ORP measurements with the HI3131B probe (sold separately)

Specifications

Includes pH electrode, temperature probe, 1m cable, pH 4.01 / 7.01 buffer solution sachets, electrode cleaning solution sachets, calibration screwdriver, battery, protective case, instructions
Accuracy (Temperature) ±0.4°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
For Use With (Application) Ideal for education and field applications
Calibration Manual via Trimmers
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) 1mV
Calibration Points 2
Temperature Range (Metric) 0°C to 100°C
Temperature Range (English) 32°F to 212°F
Resolution (Temperature) 0.1°C
Temperature Probe Calibration Slope/Offset Calibration: from 80 to 110% ±1pH
Inputs BNC connectors
Dimensions (L x W x H) 145 x 80 x 36 mm
Weight (Metric) 230 g
Weight (English) 8.1 oz.
Material Stainless Steel Temperature Probe, PEI-body pH Electrode,
Probe Type HI1230B pH Electrode and HI7669AW Temperature Probe, with BNC connector
Detection Range 0 to 14 pH
Temperature Compensation Automatic, 0 to 70°C
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Battery Type 9V
Type Analog pH/mV/Temp Meter
Battery Life 100 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.