missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-4™ Polymer for the 310 Genetic Analyzer

Product Code. 15869726 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Samples:
Up to 500 Samples/5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples unitSize
15869726 Up to 500 Samples/5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869726 Supplier Applied Biosystems™ Supplier No. 402838

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

1mL Glass Syringe, 310 Genetic Analyzer Capillary, 47cm (36cm wtr), 310 Genetic Analyzer Capillary, 61cm (50cm wtr), 310 Running Buffer, 10X, AmpFLSTR™ Yfiler™ PCR Amplification Kit, AmpFLSTR™ Identifiler™ PCR Amplification Kit, AmpFLSTR™ SGM Plus™ PCR Amplification Kit, AmpliTaq Gold™ DNA Polymerase, 1000 units with Buffer II and MgCl2 solution, BigDye™ Terminator v1.1 Cycle Sequencing Kit, GeneScan™ 400HD ROX™ Size Standard, GeneScan™ 500 LIZ™ Size Standard, GeneScan™ 500 ROX™ Size Standard, Genetic Analyzer Sample Tubes, 0.5mL, Hi-Di™ Formamide, MicroAmp™ 8-Tube Strip, 0.2mL, MicroAmp™ Reaction Tube with Cap, 0.2mL, POP-4™ Polymer for 3130/3130xl Genetic Analyzers, POP-6™ Polymer for the 310 Genetic Analyzer, Quantifiler™ Human DNA Quantification Kit, Septa for 0.5mL Sample Tubes

POP-4™is a denaturing polymer used for the 310 Genetic Analyzer.

  • Easy to Use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
  • Robust Formulation: Formulated for sequencing and fragment analysis applications
  • Polymers allow capillaries to be used multiple times
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One bottle. Store at 2°C to 8°C.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Fragment Analysis (Denaturing), Resequencing
No. of Samples Up to 500 Samples/5 mL
On Instrument Life Usage Stability on instrument 7 days, Up to 500 samples/5 ml
Precision SD <0.15 nt to 250 nt (for 36 cm capillary)
Product Line POP-4
Product Type DNA Sequencing Reagents
Quantity 5 mL
Resolution 1 base detection up to 250 nt

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.