Learn More
(±)-Pomolidomide-d3 (phthalimide-4,5,6-d3), CDN
Specifications
Specifications
| Chemical Name or Material | Pomalidomide-D3 (phthalimide-5,6,7-D3) |
| CAS | 2093128-28-8 |
| Molecular Formula | C13 D3 H8 N3 O4 |
| Quantity | 0.005 g |
| Synonym | 1 H-Isoindole-1,3(2 H)-dione-4,5,6-d3, 7-amino-2-(2,6-dioxo-3-piperidinyl)- (ACI), 7-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione-4,5,6-d3 (ACI), (±)-Pomolidomide-d3 (phthalimide-4,5,6-d3), Pomalidomide-d3, Pomalidomide-D3 (phthalimide-5,6,7-D3), 4-Amino-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione-5,6,7-d3 |
| InChI Formula | InChI=1 S/C13H11N3O4/c14-7-3-1-2-6-10(7)13(20)16(12(6)19)8-4-5-9(17)15-11(8)18/h1-3,8 H,4-5,14H2,(H,15,17,18)/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(N)c2C(=O)N(C3CCC(=O)NC3=O)C(=O)c2c1[2 H] |
| IUPAC Name | 4-amino-5,6,7-trideuterio-2-(2,6-dioxopiperidin-3-yl)isoindole-1,3-dione |
| Molecular Weight (g/mol) | 276.263 |
| Formula Weight | 276.094 g/mol |
| Show More |