missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMK/SGK196 Antibody (6F10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084197-M03
This item is not returnable.
View return policy
Description
POMK/SGK196 Monoclonal antibody specifically detects POMK/SGK196 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), KnockDown
Specifications
| POMK/SGK196 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ23356, MGC126597, protein kinase-like protein SgK196, Sugen kinase 196 | |
| FLJ23356 (NP_115613, 251 a.a. ∽ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GDFVAPEQLWPYGEDVPFHDDLMPSYDEKIDIWKIPDISSFLLGHIEGSDMVRFHLFDIHKACKSQTPSERPTAQDVLETYQKVLDTLRDAMMSQAREML | |
| 0.1 mg | |
| Protein Kinase | |
| 84197 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| 6F10 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen, Knockdown Validated | |
| NP_115613 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction