Learn More
Tocris Bioscience™ Pomalidomide 4'-alkylC8-amine
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Pomalidomide 4'-alkylC8-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC8 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-[(8-Aminooctyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride |
| CAS | 2446474-06-0 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C21H28N4O4.HCl |
| Formula Weight | 1768 |
| Purity | 95% |