Learn More
RETSCH Polystyrene Mixing Beakers
Shop All Retsch Products
Click to view available options
Capacity (Metric):
28 mL
56 mL
Unit Size:
Each
Pack of 100
Specifications
Specifications
| Product Type | Mixing Beaker |
| Capacity (Metric) | 28 mL |
| For Use With (Equipment) | For Mixer Mill MM 400 |
| Material | Polystyrene |