Learn More
Greiner Bio-One Polystyrene CELLSTAR Cell Culture Tubes
Product Code. 10003381
Click to view available options
Closure Type:
Dual-position Vent Stopper
Screw Cap
Capacity (Metric):
12 mL
14 mL
Unit Size:
Pack of 1000
Pack of 750
Product Code. 10003381
Brand: Greiner Bio-One 191160
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Improved cell adhesion due to physical surface treatment.
  • TC treated
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 18 mm
Product Type Culture Tube
Closure Type Dual-position Vent Stopper
Length (Metric) 95 mm
Material Polystyrene
Surface Treatment TC Treated
Sterility Sterile
Capacity (Metric) 14 mL
Closure Included Yes