Learn More
JG Finneran Associates™ Plastic Big Mouth Screw Thread Vials
Description
- 40% larger opening prevents broken needles due to increased target area
- ID design incorporates the StepVial feature that precisely centers a limited volume insert in the vial neck
- Polypropylene is compatible with many solvents and an excellent choice for storing pH sensitive samples
Specifications
Specifications
| Color | Translucent |
| Diameter (Metric) Outer | 12 mm |
| Material | Polypropylene |
| Volume (Metric) | 500 μL |
| For Use With (Application) | Compatible with many solvents and an excellent choice for storing pH sensitive samples |
| Neck Style | Screw |
| No. per Case | 1000/Pk. |
| No. per Pack | 100/Pk. |
| Closure Type | Screw Cap |
| Quantity | 1000/Pk. |
| Show More |