Learn More
Berlin Packaging™ Polypropylene Screw Caps
Product Code. 30408790
Click to view available options
Thread Size (Metric):
DIN 32
DIN 40
DIN 55
Unit Size:
Pack of 14800
Pack of 26000
Pack of 60000
This item is not returnable. View return policy
Product Code. 30408790
Brand: Berlin Packaging™ 1001825
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Use Berlin Packaging™ Polypropylene Screw Caps atop bottles containing analytical lab samples, especially those containing high purity chemicals, strong acids and solvents.
  • Autoclavable
  • Polypropylene
  • For use with screw-top bottles
Specifications
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Thread Size (Metric) DIN 32
Neck Finish 07 mm
Product Type Screw Cap