Learn More
Brady™ Polypropylene Prohibition Signs: Fire, Open Flame, and Smoking Prohibited
Product Code. 10557614 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
200 x 200 mm
315 x 315 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10557614 200 x 200 mm Each
10356194 315 x 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10557614 Supplier Brady™ Supplier No. 252003
  • ONLINE EXCLUSIVE PRICE
    13.60 €
    12.26 € / Pack of 1
    Save 1.34 €
    9% Off
Estimated Shipment: 17-11-2026