Learn More
Multiroir Controlec™ Polypropylene Plastic Transport Trays
Product Code. 11750594
Click to view available options
Weight (Metric):
1.6 kg
2.5 kg
3.8 kg
7 kg
Unit Size:
Each
Product Code. 11750594
Brand: Multiroir Controlec™ AMBD4322
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Transport Tray
Color Blue
Material Polypropylene
Depth (Metric) 299 mm
Depth (Metric) Interior 235 mm
Height (Metric) 240 mm
Height (Metric) Interior 198 mm
Length (Metric) 409 mm
Length (Metric) Interior 313 mm
Model AMBD4322
Weight (Metric) 1.6 kg