Learn More
Corning™ Polypropylene Pipette Tip
Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Pack of 30000
Description
Order Info
dimColor
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Color | Natural |
| Clean Claims | DNase, RNase, and Pyrogen Free |
| Material | Polypropylene |