Learn More
Fisherbrand™ Large Orifice Pipet Tips
Wide opening helps prevent damage to fragile cells and genomic DNA during pipetting. Fisherbrand™ Large Orifice Pipet Tips may also be helpful when pipetting viscous solutions.
Brand: Fisherbrand™
Additional Details : Weight : 0.05000kg
Description
- Natural color
- 200μL capacity
- For use with ErgoOne™, Eppendorf™, Biohit™, Pipetman™, USA Scientific™, Finnpipette™, Nichiryo™, Oxford Benchmate™ and other similar pipettors
Specifications
200 μL | |
Non-sterile | |
Polypropylene |