Learn More
Fisherbrand™ Large Orifice Pipet Tips
Wide opening helps prevent damage to fragile cells and genomic DNA during pipetting. Fisherbrand™ Large Orifice Pipet Tips may also be helpful when pipetting viscous solutions.
Brand:  Fisherbrand™
Additional Details : Weight : 0.05000kg
Product Code. 11582343
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Natural color
  • 200μL capacity
  • For use with ErgoOne™, Eppendorf™, Biohit™, Pipetman™, USA Scientific™, Finnpipette™, Nichiryo™, Oxford Benchmate™ and other similar pipettors
Specifications
Specifications
200 μL
Non-sterile
Polypropylene