Learn More
BrandTech™ VITLAB™ Polypropylene Pipet Fillers
Product Code. 10662631
Encompass
Click to view available options
For Use With (Equipment):
Pipettes, 10 mL
Pipettes, 25 mL
Color:
Green
Red
Unit Size:
Each
Product Code. 10662631
Brand: BrandTech™ 002524
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Designed to fit all glass and plastic pipets. VITLab™ Polypropylene Pipet Fillers are color coded for easy volume identification.
  • For pipetting liquids
  • Slow rotation of the actuator-wheels draws liquid into the pipette
  • Pressing the air bleed valve automatically empties the pipette without returning the piston
Specifications
Material Polypropylene
For Use With (Application) Pipetting liquids
Quantity 1 Each
Color Red
Type Pipette Filler
For Use With (Equipment) Pipettes, 25 mL