Learn More
Greiner Bio-One Polypropylene Micro Pipet Tips
Designed for precise pipetting of the smallest possible liquid volumes. Greiner Bio-One™ Polypropylene Micro Pipet Tips provide a small dead volume due to their small size.
Brand:  Greiner Bio-One 771289
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10569822
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Manufactured from high-grade polypropylene for optimum performance and fit
  • High breaking strength, dimensionally stable
  • Heat resistant to +140°C 
  • Free of heavy metals
  • Available in bulk pacaging or convenient rack
  • Conform to ISO 9001 standard
Specifications
Specifications
0.5 to 10 μL
Natural
P10 standard
Gilson™ Pipetman™ U10, Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2
Sterile
Autoclavable
4800/Pack
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers