Learn More
VITLAB™ Polypropylene Measuring Pipets
Specifications
Specifications
| Material | Polypropylene |
| Graduated | Yes |
| Graduations | 0.1 mL |
| Quantity | 1 Pack |
| Tolerance | ±0.10 mL |
| Volume (Metric) | 10 mL |
| Length (Metric) | 330 mm |
| Type | Measuring Pipet |