Learn More
Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes
Product Code. 11568242
Change view
Click to view available options
Color:
Blue
Natural
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11568242 Blue Pack of 500
10604555 Natural Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11568242 Supplier Fisherbrand™ Supplier No. T33110B
Estimated Shipment: 21-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Use these tubes for a variety of applications. Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes are ideal for use with PCR systems and centrifugation.
  • Polypropylene graduated tubes
  • Autoclavable
  • Available in a variety of colors for easy identification
  • Available with pick-up tab
Specifications
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Blue
Material Polypropylene
For Use With (Application) For PCR, Centrifugation
No. per Pack 500/Pk.
Graduated Yes
Closure Type Snap Cap