Learn More
Hirschmann™ Polypropylene Disposable Microplate
Beschreibung
Flat bottom not treated disposable plastic with fixed glass inserts clear 1200μL 96 well Hirschmann
Spezifikation
Spezifikation
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Disposable | Single-use |
| Product Type | Microplate |
| Material | Polypropylene |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 1200 μL |
| Well Shape | Flat Bottom (F-well) |
| No. per Pack | 1 Each |
| Mehr anzeigen |