missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Polypropylene, Deep Freeze Box

Product Code. 10711624
Encompass
Click to view available options
Dimensions (L x W x H):
144 x 144 x 127 mm
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 10711624

Brand: Carl Roth™ HX02.1

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Specifications

Closure Included Slip Lid
Autoclavable Autoclavable
Color Blue
Dimensions (L x W x H) 144 x 144 x 127 mm
Height (Metric) 144 mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.