Learn More
Masterflex™ Polypropylene Barbed Fitting
Description
- Quick and easy assembly
- Available in packs of ten
Specifications
Specifications
| Material | Polypropylene |
| For Use With | 3/8 NPT x 3/8 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Port Size | 1/4 in. orifice; 1-9/16 x 1-19/32 in. |
| Thread Size | 3/8 in. NPT |
| Shape | Elbow |
| Includes | ETO or gamma sterilization |
| Color | Natural |
| Product Type | Barbed Fitting |