Learn More
Masterflex™ Polypropylene Barbed Fitting
Product Code. 12147770
Change view
Click to view available options
For Use With:
1/2 NPT x 1/2 in. tubing I.D.
1/2 NPT x 1/4 in. tubing I.D.
1/4 NPT x 1/2 in. tubing I.D.
1/4 NPT x 3/8 in. tubing I.D.
3/8 NPT x 1/2 in. tubing I.D.
3/8 NPT x 1/4 in. tubing I.D.
3/8 NPT x 3/8 in. tubing I.D.
Shape:
Elbow
Straight
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Shape unitSize
12147770 1/4 NPT x 1/2 in. tubing I.D. Straight Pack of 10
11979658 1/2 NPT x 1/2 in. tubing I.D. Straight Pack of 10
13263789 1/2 NPT x 1/4 in. tubing I.D. Straight Pack of 10
13294869 1/2 NPT x 1/2 in. tubing I.D. Elbow Pack of 10
12636776 1/4 NPT x 3/8 in. tubing I.D. Straight Pack of 10
11740329 3/8 NPT x 1/4 in. tubing I.D. Elbow Pack of 10
12557376 3/8 NPT x 3/8 in. tubing I.D. Elbow Pack of 10
11750329 3/8 NPT x 1/2 in. tubing I.D. Elbow Pack of 10
11989628 3/8 NPT x 3/8 in. tubing I.D. Straight Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12147770 Supplier Masterflex™ Supplier No. 4061040
Only null left
Add to basket
Add to basket
Adapts from a NPT male thread to a hose barb. Cole-Parmer™ Polypropylene Barbed Fittings are available in a variety of configurations to meet your needs.
  • Quick and easy assembly
  • Available in packs of ten
Specifications
Material Polypropylene
For Use With 1/4 NPT x 1/2 in. tubing I.D.
Packaging Quantity 10 Pack
Port Size 11/32 in. orifice; 1-29/32 x 21/32 in.
Thread Size 1/4 in. NPT
Shape Straight
Includes ETO or gamma sterilization
Product Type Barbed Fitting