Learn More
Buerkle™ Polypropylene Ball Valve, Food
Product Code. 10204744 Shop All Bürkle Products
Click to view available options
Width (Metric):
20 mm
25 mm
N/A
Outlet Port:
M29 x 1,75, Outer
M36 x 3, Outer
M42 x 3, Outer
N/A
Unit Size:
Each
Product Code. 10204744
Brand: Buerkle™ 05008203
Only null left
Add to basket
Add to basket
Ball valve made of PP with outer / inner thread, fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and seal made of HDPE.
  • Max. operating pressure 1 bar (at +20 °C)
  • Max. operating temperature 100 °C
  • Suitable for food
TRUSTED_SUSTAINABILITY
Specifications
Type Ball Valve
Material Polypropylene
Pressure 10 bar
Width (Metric) 20 mm
Includes Outer/inner thread, fibre-Glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and seal made of HDPE
Outlet Port N/A