Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Product Code. 10610713
Click to view available options
Thread Size:
GL 25
GL 28
GL 30
GL 33
GL 36
GL 55
GL 60
GL 65
Unit Size:
Each
Product Code. 10610713
Brand: Poulten & Graf GmbH 101 09025
Only null left
Add to basket
Add to basket
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with threads
  • Can be used for most chemicals and liquids
Specifications
Product Type Adapters for OPTIFIX and POLYFIX Dispensers
Autoclavable Autoclavable
Color White
Description Adapters for OPTIFIX and POLYFIX dispensers
Material Polypropylene
For Use With (Equipment) OPTIFIX and POLYFIX dispensers
Quantity 1
Thread Size GL 25