Learn More
Macherey-Nagel™ Polygoprep™ 300 to 20 C4
Product Code. 10184831 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
0.1 kg
1 kg
Unit Size:
1000g
100g
Product Code. 10184831
Brand: Macherey-Nagel™ 711023.100
Please to purchase this item. Need a web account? Register with us today!
Specifications
Particle Size 15 to 25 μm
Pore Size 300 Å
Stationary Phase POLYGOPREP 300 to 20
pH 2 to 8
Quantity 0.1 kg
Sorbent POLYGOPREP C4
Type Polygoprep
For Use With (Application) Liquid Chromatography