Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30
Product Code. p-8012348 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
0.1 kg
1 kg
Particle Size:
10 to 15 μm
25 to 40 μm
40 to 63 μm
Unit Size:
1000g
100g
Product Code. 10082711
Brand: Macherey-Nagel™ 711005.100
Only null left
Add to basket
Add to basket
Specifications
Particle Size 10 to 15 μm
Pore Size 1000 Å
Stationary Phase POLYGOPREP 1000 to 30
pH 2 to 8
Quantity 0.1 kg
Sorbent 'POLYGOPREP
Type Polygoprep
For Use With (Application) Liquid Chromatography