Learn More
Hirschmann™ Polyethylene Poly Stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
1.50 € - 3.95 €
Specifications
Material | Polyethylene |
---|---|
Product Type | Poly Stopper |
Shape | Hexagonal |
Sterility | Non-sterile |
Quantity | 1 Each |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
10054301
|
Hirschmann™
8900252 |
Stopper Number: NS 7/16 |
1.50 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10125261
|
Hirschmann™
8900260 |
Stopper Number: NS 10/19 |
1.50 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10369991
|
Hirschmann™
8900265 |
Stopper Number: NS 12/21 |
1.65 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10105311
|
Hirschmann™
8900270 |
Stopper Number: NS 14/23 |
1.80 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10640992
|
Hirschmann™
8900293 |
Stopper Number: NS 45/40 |
3.95 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- PE
- Available to fit a range of sizes
Specifications
Polyethylene | |
Hexagonal | |
1 Each |
Poly Stopper | |
Non-sterile |