Learn More
Brady™ Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Product Code. 12674883
Click to view available options
Color:
White
White, Transparent
Width (Metric):
9.53 mm
15.24 mm
25.4 mm
48.26 mm
Height (Metric):
9.53 mm
12.7 mm
15 mm
25.40 mm
38.1 mm
41.28 mm
44.45 mm
45.72 mm
66.68 mm
Unit Size:
Each
Product Code. 12674883
Brand: Brady™ 710781
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Low-profile label for use on frozen surfaces or on room-temperature surfaces.
  • Opaque to cover legally protected patient information
  • Ultra thin
  • Lasts in liquid nitrogen, autoclaves and hot water baths
  • Permanent Acrylic adhesive
Specifications
Description BMP71 FreezerBondz Label Printer Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Material Polyester
Print Technology Thermal Transfer
Product Type Label Printer Label
For Use With (Application) Laboratory identification
For Use With (Equipment) BMP™71
Labels per Row 1
No. per Pack 300/Pk.
Quantity 300 Pack
Shape Rectangle
Size (Metric) 48.26W x 25.4 mmH
Thickness (Metric) 0.020 mm
Vertical Repeat (Metric) 28.58 mm
Dimensions (LxW) 25.40 x 48.26 mm
Width (Metric) 48.26 mm
Height (Metric) 25.40 mm