Learn More
Brady™ Polyester Mandatory Safety Glasses Signs
Product Code. 11754964
Click to view available options
Unit Size:
Each
Product Code. 11754964
Brand: Brady™ 806847
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance RoHS compliant using EU Directive 2002/95/EC
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
For Use With (Application) Pipe markers and safety signs
Height (Metric) 105 mm
Width (Metric) 297 mm
Shape Rectangular
Displayed Text Safety glasses
Product Type Mandatory Sign
Color White