Learn More
Brady™ Polyester Labels for the BBP33/i3300 Printer
Product Code. 17110521
Click to view available options
Color:
Blue, White
Green, White
Orange, White
Pink, White
Red, White
White
Yellow, White
Length (Metric):
9.53 mm
12.70 mm
15.24 mm
Unit Size:
Pack of 1500
Pack of 2500
Product Code. 17110521
Brand: Brady™ 307001
Only null left
Add to basket
Add to basket
Our B-494 polyester label is a reliable sample identification and traceability solution.
  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
  • Thanks to subsurface printing, the label's color code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Yellow, White
Finish Gloss
Format Roll
Material Polyester
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Cold temperature labelling, Component and equipment labelling, Laboratory labelling
For Use With (Equipment) BBP™ 33 Label Printer, BradyPrinter i3300 Industrial Label Printer
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
No. per Pack 2500 Labels/Box
Quantity 2500 Labels/Roll
Shape Rectangle
Temperature Range (English) -320.8°F to +158°F
Temperature Range (Metric) -196°C to +70°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.0762 mm
Vertical Repeat (Metric) 15.24 mm
Dimensions (LxW) 12.70 x 25.4 mm
Length (English) 0.5 in.
Length (Metric) 12.70 mm
Width (English) 1 in.
Width (Metric) 25.40 mm
Height (English) 0.5 in.
Height (Metric) 12.70 mm
Alternate Part Number B33-310-494-YL