Learn More
Macherey-Nagel™ Polyamide CC 6
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 kg
5 kg
Particle Size:
<0.07
0.05 to 0.16 mm
0.10 to 0.3 mm
Unit Size:
1kg
5kg
Specifications
Specifications
| Particle Size | 0.05 to 0.16 mm |
| Quantity | 1 kg |
| Type | Polyamide CC 6 |
| Material | Polyamide |