Learn More
JG Finneran Associates™ Poly Crimp™ Seals
Product Code. 10025570
Click to view available options
Material (Liner):
PTFE
PTFE/Silicone with Slit
PTFE/Silicone with Starburst
PTFE/Silicone/PTFE
Unit Size:
Pack of 1000
Product Code. 10025570
Brand: JG Finneran Associates™ 557011
Please to purchase this item. Need a web account? Register with us today!
Specifications
Material (Liner) PTFE/Silicone with Slit
Color Clear
Material PTFE, Silicone
Quantity 1000/Pk.
Closure Size 11 mm
For Use With (Equipment) 12 x 32 mm Sample Vials with Crimp Style Finish