missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2E Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene
Description
POLR2E Monoclonal antibody specifically detects POLR2E in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
| Antigen | POLR2E |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI2A11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | UBXN2B |
| Gene Accession No. | Q14CS0 |
| Gene Alias | 2410021N14Rik; 25kDa; AW208866; DNA directed RNA polymerase II 23 kda polypeptide; DNA-directed RNA polymerase II 23 kDa polypeptide; DNA-directed RNA polymerase II subunit E; DNA-directed RNA polymerase subunit RPABC1; DNA-directed RNA polymerases I, II, and III subunit RPABC1; hRPB25; hsRPB5; Polr2e; polymerase (RNA) II (DNA directed) polypeptide E; polymerase (RNA) II (DNA directed) polypeptide E, 25kDa; polymerase (RNA) II subunit E; RNA polymerase II subunit E; RNA polymerase II, I and III subunit E; RNA polymerases I, II, and III subunit ABC1; RPABC1; RPB5; RPB5 homolog; XAP4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?