Learn More
POLR2E Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502490
Encompass_Preferred
 View more versions of this product
Product Code. 17517031
  • 540.87 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
This gene encodes the fifth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit is shared by the other two DNA-directed RNA polymerases and is present in two-fold molar excess over the other polymerase subunits. An interaction between this subunit and a hepatitis virus transactivating protein has been demonstrated, suggesting that interaction between transcriptional activators and the polymerase can occur through this subunit. A pseudogene is located on chromosome 11.
Specifications
Specifications
POLR2E
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P19388
POLR2E
Full length human recombit protein of human POLR2E produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A11
Unconjugated
POLR2E
RPABC1, RPB5, XAP4, hRPB25, hsRPB5
Mouse
Affinity Chromatography
RUO
5434
-20° C, Avoid Freeze/Thaw Cycles